Search
Search

Kctd14| Gene Symbol: Kctd14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001010826.3 | NP_001010826.2 | 3 | 405 | ||
| NM_001012434.3 | NP_001012436.1 | 3 | 438 | |||
| NM_001136235.1 | NP_001129707.1 | 3 | 422 | |||
| GenBank mRNA | AK145279.1 | - | 3 | 406 | ||
| AY899809.1 | - | 3 | 438 | |||
| BC024588.1 | - | 3 | 277 | 5066294 | ||
| BG088171.2 | - | 3 | 422 | |||
| BY735425.1 | - | 3 | 406 | |||
| CK791017.1 | - | 3 | 326 | |||