Search
Search

Ppfia1| Gene Symbol: Ppfia1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033319.2 | NP_001028491.1 | 6 | 887 | ||
| NM_001195086.1 | NP_001182015.1 | 6 | 887 | |||
| GenBank mRNA | BC157936.1 | - | 6 | 704 | 9088116 | |
| BC158007.1 | - | 6 | 844 | 9007315 | ||
| BC172096.1 | - | 6 | 707 | 9054382 | ||
| EU568869.1 | - | 6 | 887 | |||