Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Rltpr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033320.2 | NP_001028492.1 | 12, 13 | 1951 | ||
GenBank mRNA | AK031012.1 | - | 12, 13 | 1950 | ||
HF678090.1 | - | 33, 34 | 3582 |