Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Insl5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290648.1 | NP_001277577.1 | 2 | 132 | ||
NM_011831.2 | NP_035961.1 | 1 | 138 | |||
GenBank mRNA | AF054842.1 | - | 2 | 132 | ||
AF054843.1 | - | 1 | 102 | |||
AF076971.1 | - | 1 | 134 | |||
AF133817.1 | - | 1 | 141 | |||
BC010968.1 | - | 1 | 133 | 4217383 |