Search Thermo Fisher Scientific
- ์ฃผ๋ฌธ ํํฉ
- ๋น ๋ฅธ์ฃผ๋ฌธ
-
ํ์์ด ์๋์ ๊ฐ์? ๊ณ์ ์์ฑํ๊ธฐ
Search Thermo Fisher Scientific
Gene Symbol: Abce1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015751.2 | NP_056566.2 | 7 | 783 | ||
GenBank mRNA | AK049299.1 | - | 7 | 784 | ||
AK133748.1 | - | 7 | 714 | |||
AK134738.1 | - | 7 | 720 | |||
AK144711.1 | - | 7 | 712 | |||
AK147151.1 | - | 7 | 721 | |||
AK167130.1 | - | 7 | 731 | |||
AK167370.1 | - | 7 | 721 | |||
AK167704.1 | - | 7 | 682 | |||
BC005422.1 | - | 7 | 704 | 3489529 | ||
BC066794.1 | - | 6 | 486 | |||
BC066836.1 | - | 6 | 486 | |||
U90446.1 | - | 7 | 672 |