Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mpdu1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301710.1 | NP_001288639.1 | 4 | 426 | ||
NM_001301711.1 | NP_001288640.1 | 6 | 666 | |||
NM_011900.4 | NP_036030.2 | 6 | 631 | |||
GenBank mRNA | AB024713.1 | - | 6 | 559 | ||
AK040432.1 | - | 6 | 570 | |||
AK080233.1 | - | 6 | 625 | |||
AK089426.1 | - | 6 | 573 | |||
AK172020.1 | - | 6 | 592 | |||
AK188244.1 | - | 1 | 22 | |||
BC026776.1 | - | 6 | 553 | 4159355 | ||
BC048871.1 | - | 6 | 559 | 5364408 | ||
CV561188.1 | - | 6 | 617 | |||
CV561394.1 | - | 5 | 465 | |||
W67006.1 | - | 2 | 133 |