Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Dhrs9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175512.2 | NP_780721.1 | 2 | 342 | ||
GenBank mRNA | AF361479.1 | - | 1 | 127 | ||
AK050180.1 | - | 2 | 360 | |||
AK080914.1 | - | 2 | 342 | |||
AY936479.1 | - | 1 | 114 | |||
BC118029.1 | - | 2 | 303 | 40093681 |