Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Rgs9bp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145840.3 | NP_665839.1 | 1 | 629 | ||
GenBank mRNA | AF483907.1 | - | 1 | 267 | ||
AY229888.1 | - | 1 | 591 | |||
BC118006.1 | - | 1 | 585 | 40081388 |