Search Thermo Fisher Scientific
Gene Symbol: Rgs9bp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145840.3 | NP_665839.1 | 1 | 953 | ||
GenBank mRNA | AF483907.1 | - | 1 | 591 | ||
AK044285.1 | - | 1 | 954 | |||
AY229888.1 | - | 1 | 915 | |||
BC118006.1 | - | 1 | 909 | 40081388 |