Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rbbp7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009031.3 | NP_033057.3 | 5 | 897 | ||
GenBank mRNA | AK076016.1 | - | 5 | 822 | ||
AK078349.1 | - | 5 | 810 | |||
AK082892.1 | - | 5 | 809 | |||
AK132101.1 | - | 5 | 808 | |||
AK135779.1 | - | 5 | 811 | |||
AK135869.1 | - | 4 | 506 | |||
AK135875.1 | - | 4 | 506 | |||
AK135956.1 | - | 5 | 811 | |||
AK136110.1 | - | 5 | 809 | |||
AK145531.1 | - | 5 | 808 | |||
AK145651.1 | - | 5 | 791 | |||
AK146014.1 | - | 5 | 809 | |||
AK146439.1 | - | 5 | 868 | |||
AK146904.1 | - | 5 | 810 | |||
AK146967.1 | - | 5 | 809 | |||
AK147062.1 | - | 5 | 868 | |||
AK148852.1 | - | 5 | 837 | |||
AK153913.1 | - | 5 | 854 | |||
AK160023.1 | - | 5 | 809 | |||
BC003785.1 | - | 5 | 826 | 3600013 | ||
U35142.1 | - | 5 | 836 |