Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Olfr1273-ps | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146975.1 | NP_667186.1 | 1 | 601 | ||
GenBank mRNA | BC104266.1 | - | 1 | 601 | 40040402 | |
BC104267.1 | - | 1 | 601 | 40040403 | ||
BC126917.1 | - | 1 | 601 | 40040394 |