Search
Search

Gdpgp1| Gene Symbol: Gdpgp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178752.3 | NP_848867.2 | 2 | 1246 | ||
| GenBank mRNA | AK084531.1 | - | 2 | 1232 | ||
| AK085577.1 | - | 1254 | ||||
| AK086166.1 | - | 2 | 1249 | |||
| AK143704.1 | - | 2 | 1272 | |||