Search
Search

Tmcc3| Gene Symbol: Tmcc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001168684.1 | NP_001162155.1 | 2 | 755 | ||
| NM_172051.3 | NP_742048.2 | 2 | 832 | |||
| NM_177026.1 | NP_796000.1 | 2 | 774 | |||
| GenBank mRNA | AK079611.1 | - | 3 | 692 | ||
| AK083129.1 | - | 2 | 735 | |||
| AK132545.1 | - | 2 | 810 | |||
| BC026867.1 | - | 2 | 755 | 3660220 | ||
| BC057202.1 | - | 2 | 618 | 5327878 | ||
| BC095951.1 | - | 2 | 766 | 5034018 | ||