Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Cass4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080820.2 | NP_001074289.1 | 5 | 1576 | ||
NM_001276422.1 | NP_001263351.1 | 5 | 1582 | |||
NM_001276423.1 | NP_001263352.1 | 5 | 1576 | |||
GenBank mRNA | AK036419.1 | - | 5 | 1550 | ||
AK080960.1 | - | 5 | 1551 | |||
AK089450.1 | - | 5 | 1577 | |||
AK170726.1 | - | 5 | 1576 | |||
BC117858.1 | - | 5 | 1470 | 40095182 | ||
BC117859.1 | - | 5 | 1469 | 40095183 | ||
BC129975.1 | - | 5 | 1467 | 40095184 | ||
BC129976.1 | - | 5 | 1475 | 40095185 |