Search
Search

Fcrl5| Gene Symbol: Fcrl5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001113238.1 | NP_001106709.1 | 6 | 1224 | ||
| NM_183222.3 | NP_899045.3 | 7 | 1488 | |||
| GenBank mRNA | AK089756.1 | - | 6 | 1194 | ||
| AY158090.1 | - | 7 | 1485 | |||
| AY506558.1 | - | 7 | 1494 | |||
| BC137997.1 | - | 7 | 1422 | 8861016 | ||
| BC137998.1 | - | 7 | 1422 | 8861017 | ||