Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tceal5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177919.2 | NP_808587.1 | 3 | 765 | ||
GenBank mRNA | AK032140.1 | - | 3 | 767 | ||
BC100415.1 | - | 3 | 790 | 6535676 | ||
BC115460.1 | - | 2 | 676 | 40100235 | ||
BC115461.1 | - | 2 | 676 | 40100237 |