Search
Search

H2-M10.2| Gene Symbol: H2-M10.2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177923.1 | NP_808591.1 | 2 | 299 | ||
| GenBank mRNA | AF539598.1 | - | 2 | 299 | ||
| AY211078.1 | - | 282 | ||||
| BC137985.1 | - | 2 | 368 | 8861004 | ||
| BC137987.1 | - | 2 | 368 | 8861006 | ||