Search Thermo Fisher Scientific
- ์ฃผ๋ฌธ ํํฉ
- ๋น ๋ฅธ์ฃผ๋ฌธ
-
ํ์์ด ์๋์ ๊ฐ์? ๊ณ์ ์์ฑํ๊ธฐ
Search Thermo Fisher Scientific
Gene Symbol: Ano8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164679.1 | NP_001158151.1 | 13 | 2397 | ||
GenBank mRNA | AK173222.1 | - | 11 | 3385 | ||
BC027735.1 | - | 13 | 2299 | |||
BC027790.1 | - | 6 | 1871 | |||
BC042391.1 | - | 6 | 2592 | |||
BC053460.1 | - | 1 | 436 | |||
BC059855.1 | - | 4 | 930 |