Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Ano8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164679.1 | NP_001158151.1 | 15, 16 | 2749 | ||
GenBank mRNA | AK173222.1 | - | 13, 14 | 3737 | ||
BC027735.1 | - | 15, 16 | 2651 | |||
BC042391.1 | - | 8, 9 | 2944 | |||
BC053460.1 | - | 3, 4 | 788 | |||
BC059855.1 | - | 6, 7 | 1282 |