Search
Search

Rhpn2| Gene Symbol: Rhpn2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027897.4 | NP_082173.3 | 14 | 1770 | ||
| GenBank mRNA | AK004849.1 | - | 14 | 1693 | ||
| AK050214.1 | - | 14 | 1684 | |||
| BC100314.1 | - | 14 | 1677 | 30848276 | ||