Search
Search

Nucb2| Gene Symbol: Nucb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001130479.2 | NP_001123951.1 | 6, 7 | 779 | ||
| GenBank mRNA | AJ222586.1 | - | 6, 7 | 779 | ||
| AK145934.1 | - | 6, 7 | 734 | |||
| AK152672.1 | - | 17 | ||||
| AK160442.1 | - | 5, 6 | 627 | |||
| BC010459.1 | - | 6, 7 | 693 | 3585076 | ||