Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Uevld | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040695.1 | NP_001035785.1 | 8 | 890 | ||
GenBank mRNA | AK154536.1 | - | 8 | 841 | ||
AK155685.1 | - | 8 | 837 | |||
BC054796.1 | - | 8 | 870 | 6843964 |