Search
Search

Gabarap| Gene Symbol: Gabarap | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019749.4 | NP_062723.1 | 2 | 463 | ||
| GenBank mRNA | AF161587.1 | - | 2 | 183 | ||
| AK002879.1 | - | 2 | 218 | |||
| AK011731.1 | - | 2 | 128 | |||
| AV029091.2 | - | 2 | 234 | |||
| BC002126.1 | - | 2 | 226 | 3497484 | ||
| BC024621.1 | - | 2 | 246 | 4459651 | ||
| BC029329.1 | - | 2 | 165 | |||
| BC030350.1 | - | 2 | 472 | 5102114 | ||