Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Trim17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031172.2 | NP_112449.1 | 7 | 1455 | ||
GenBank mRNA | AF220135.1 | - | 1 | 215 | ||
AK077212.1 | - | 7 | 1438 | |||
BC055112.1 | - | 7 | 1455 | 6707359 |