Search
Search

Tas2r119| Gene Symbol: Tas2r119 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020503.2 | NP_065249.2 | 1 | 838 | ||
| GenBank mRNA | BC111811.1 | - | 1 | 839 | ||
| BC128078.1 | - | 1 | 835 | |||
| BC128079.1 | - | 1 | 839 | |||
| BC152359.1 | - | 1 | 839 | 40101839 | ||