Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Fam13a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153574.2 | NP_705802.1 | 9 | 1060 | ||
GenBank mRNA | AJ510136.1 | - | 9 | 1046 | ||
AK036291.1 | - | 2 | 267 | |||
AK052421.1 | - | 9 | 1001 | |||
BC005554.1 | - | 9 | 965 | |||
BC054053.1 | - | 9 | 1034 | |||
BC060978.1 | - | 9 | 997 | |||
BC080861.1 | - | 9 | 1060 | 30357292 |