Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Fam71a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109759.1 | NP_001103229.1 | 1 | 754 | ||
GenBank mRNA | AB473728.1 | - | 1 | 381 | ||
AK133412.1 | - | 1 | 756 |