Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Aadacl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001128091.1 | NP_001121563.1 | 5 | 1350 | ||
GenBank mRNA | BC147724.1 | - | 5 | 1350 | 9056673 | |
BC147728.1 | - | 5 | 1350 | 9056677 |