Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Prl2b1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025532.3 | NP_079808.1 | 4 | 452 | ||
GenBank mRNA | AF525155.1 | - | 4 | 403 | ||
AK005411.1 | - | 4 | 453 | |||
AK009863.1 | - | 4 | 452 | |||
BC100329.1 | - | 4 | 442 | 30787440 | ||
BC111112.1 | - | 4 | 448 | 40050224 | ||
BC138199.1 | - | 4 | 445 | 8861218 |