Search
Search

Sys1| Gene Symbol: Sys1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025575.3 | NP_079851.2 | 2 | 292 | ||
| GenBank mRNA | AK003071.1 | - | 2 | 285 | ||
| AK005249.1 | - | 2 | 195 | |||
| AK005510.1 | - | 2 | 229 | |||
| AK007882.1 | - | 2 | 293 | |||
| AK009684.1 | - | 2 | 256 | |||
| AK011753.1 | - | 2 | 288 | |||
| AK013220.1 | - | 2 | 267 | |||
| AK017524.1 | - | 1 | 125 | |||
| AK032073.1 | - | 2 | 263 | |||
| AK032098.1 | - | 1 | 127 | |||
| AK075770.1 | - | 2 | 257 | |||
| AK139605.1 | - | 1 | 463 | |||
| AK148305.1 | - | 2 | 230 | |||
| BC038166.1 | - | 3 | 578 | |||
| BC053943.1 | - | 1 | 126 | |||
| BC054055.1 | - | 3 | 750 | |||
| BC061084.1 | - | 1 | 166 | 6596164 | ||