Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Mrgpra9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001288801.1 | NP_001275730.1 | 2 | 799 | ||
GenBank mRNA | DQ386867.1 | - | 1 | 746 |
Set Membership: |
G-protein coupled receptor |