Search
Search

Kctd20| Gene Symbol: Kctd20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025888.5 | NP_080164.2 | 4, 5 | 675 | ||
| GenBank mRNA | AK010404.1 | - | 3, 4 | 438 | ||
| AK030202.1 | - | 4, 5 | 677 | |||
| AK088807.1 | - | 3, 4 | 443 | |||
| AK168440.1 | - | 4, 5 | 666 | |||
| AK171742.1 | - | 3, 4 | 473 | |||
| BC036318.1 | - | 2, 3 | 320 | |||
| BC048161.1 | - | 3, 4 | 459 | |||
| BC049250.1 | - | 3, 4 | 552 | 5042309 | ||
| BC052712.1 | - | 3, 4 | 461 | |||