Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp787 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013012.1 | NP_001013030.1 | 3 | 374 | ||
GenBank mRNA | AK077458.1 | - | 3 | 341 | ||
AK159396.1 | - | 2 | 199 | |||
BC038658.1 | - | 3 | 349 | |||
BC137753.1 | - | 3 | 296 | 8860772 |