Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Wdr83 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026399.2 | NP_080675.2 | 3, 4 | 470 | ||
GenBank mRNA | AK005790.1 | - | 3, 4 | 410 | ||
AK020654.1 | - | 3, 4 | 431 | |||
AY365434.1 | - | 3, 4 | 470 | |||
BC019369.1 | - | 3, 4 | 393 | 4014928 |