Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Snhg17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK007971.1 | - | 3 | 288 | ||
AK040893.1 | - | 3 | 279 | |||
AK046044.1 | - | 3 | 388 | |||
AK079109.1 | - | 3 | 288 |