Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Ms4a6d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026835.2 | NP_081111.1 | 5 | 582 | ||
GenBank mRNA | AF237911.1 | - | 5 | 544 | ||
AK004295.1 | - | 5 | 565 | |||
AK152859.1 | - | 5 | 562 | |||
AK156878.1 | - | 5 | 565 | |||
AY039037.1 | - | 5 | 544 | |||
BC018331.1 | - | 5 | 519 | 3964037 | ||
BC029738.1 | - | 5 | 526 | 4016611 | ||
BG087418.2 | - | 5 | 582 |