Search
Search

Gsdmd| Gene Symbol: Gsdmd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026960.4 | NP_081236.1 | 9 | 1218 | ||
| GenBank mRNA | AK007710.1 | - | 9 | 1219 | ||
| AK088747.1 | - | 9 | 1230 | |||
| AK165858.1 | - | 9 | 1219 | |||
| AK171294.1 | - | 9 | 1220 | |||
| BC029813.1 | - | 9 | 1171 | 4194837 | ||