Search
Search

Rpain| Gene Symbol: Rpain | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001252413.1 | NP_001239342.1 | 4 | 702 | ||
| NM_001252415.1 | NP_001239344.1 | 4 | 702 | |||
| NM_027186.2 | NP_081462.1 | 4 | 702 | |||
| GenBank mRNA | AK010289.1 | - | 4 | 431 | ||
| BC047996.1 | - | 4 | 689 | 6509910 | ||
| DQ810267.1 | - | 4 | 395 | |||
| DQ810268.1 | - | 4 | 395 | |||
| DQ810269.1 | - | 4 | 395 | |||