Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Slco6d1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164233.1 | NP_001157705.1 | 2 | 441 | ||
NM_027584.1 | NP_081860.1 | 2 | 441 | |||
GenBank mRNA | AK014872.1 | - | 2 | 442 | ||
BC112423.1 | - | 2 | 411 | 40062055 | ||
BC113131.1 | - | 2 | 411 | 40062053 | ||
BY714392.1 | - | 2 | 442 |