Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Slc41a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037493.2 | NP_001032570.1 | 9 | 1332 | ||
NM_027868.2 | NP_082144.2 | 10 | 1364 | |||
GenBank mRNA | AK003140.1 | - | 9 | 1332 | ||
BC011108.1 | - | 10 | 1280 | 4011836 |