Search
Search

Serpina3a| Gene Symbol: Serpina3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001167705.1 | NP_001161177.1 | 5 | 1332 | ||
| NM_028740.2 | NP_083016.1 | 4 | 672 | |||
| GenBank mRNA | AK016702.1 | - | 4 | 675 | ||
| BC063325.1 | - | 5 | 1522 | 30280343 | ||