Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gsdmc4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028992.1 | NP_083268.1 | 6, 7 | 860 | ||
GenBank mRNA | AK018537.1 | - | 6, 7 | 861 | ||
AK163061.1 | - | 6, 7 | 861 |