Search
Search

Tmem65| Gene Symbol: Tmem65 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175212.4 | NP_780421.2 | 2, 3 | 628 | ||
| GenBank mRNA | AK015335.1 | - | 2, 3 | 726 | ||
| AK031886.1 | - | 2, 3 | 653 | |||
| BC049380.1 | - | 2, 3 | 179 | 4503473 | ||
| BC096426.1 | - | 2, 3 | 628 | 2615937 | ||