Search
Search

Cmtm2b| Gene Symbol: Cmtm2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028524.3 | NP_082800.1 | 3 | 470 | ||
| GenBank mRNA | AK005970.1 | - | 3 | 464 | ||
| AY162282.1 | - | 3 | 350 | |||
| AY241872.1 | - | 3 | 464 | |||
| BC050983.1 | - | 3 | 467 | |||
| BC060997.1 | - | 3 | 363 | 6742828 | ||