Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Spopl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001165997.1 | NP_001159469.1 | 4 | 547 | ||
NM_001165998.1 | NP_001159470.1 | 3 | 411 | |||
NM_029773.2 | NP_084049.2 | 4 | 547 | |||
GenBank mRNA | AK162056.1 | - | 3 | 264 | ||
BC111867.1 | - | 4 | 421 | 40056321 | ||
BI648825.1 | - | 4 | 497 |