Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Crisp4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030033.1 | NP_084309.1 | 5, 6 | 548 | ||
GenBank mRNA | AK020340.1 | - | 6, 7 | 609 | ||
AY705449.1 | - | 5, 6 | 548 | |||
DQ200908.1 | - | 7, 8 | 725 |