Search Thermo Fisher Scientific
Gene Symbol: Golga2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080968.1 | NP_001074437.1 | 14 | 1143 | ||
NM_133852.2 | NP_598613.2 | 15 | 1224 | |||
GenBank mRNA | AK076370.1 | - | 9 | 842 | ||
AK077516.1 | - | 14 | 1145 | |||
AK220509.1 | - | 15 | 1152 | |||
BC011407.1 | - | 13 | 944 | 3596079 |