Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Enterprise Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other MAGI2 SIRNA Assays ›
Gene Symbol:
Magi2
Gene Name:
membrane associated guanylate kinase, WW and PDZ domain containing 2
Gene Aliases:
Acvrinp1, Acvrip1, S-SCAM
Chromosome Location:
Chr.4: 10995241 - 12472423 on Build Rnor_6.0
UniGene ID:
Rn.205058
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID
RSS300048
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1330001
Price
Contact Us
Your Price
Online offer:
3062.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Magi2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_053621.1
NP_446073.1
10
2134
GenBank mRNA
AF034863.1
-
10
2134
AF130819.1
-
9
1921
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
positive regulation of receptor internalization
glomerular filtration
signal transduction
negative regulation of cell proliferation
positive regulation of neuron projection development
phosphorylation
cytosolic transport
negative regulation of cell migration
positive regulation of phosphoprotein phosphatase activity
negative regulation of activin receptor signaling pathway
nerve growth factor signaling pathway
receptor clustering
protein heterooligomerization
negative regulation of protein kinase B signaling
SMAD protein signal transduction
mitotic cell cycle arrest
glomerular visceral epithelial cell development
cellular response to nerve growth factor stimulus
signal transducer activity
protein binding
kinase activity
phosphatase binding
receptor signaling complex scaffold activity
PDZ domain binding
beta-1 adrenergic receptor binding
protein complex binding
SMAD binding
type II activin receptor binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-9t45z:80/100.66.79.31:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0