Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Pou2f2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006228368.2 | XP_006228430.2 | 6, 7 | 469 | ||
XM_006228371.2 | XP_006228433.2 | 6, 7 | 535 | |||
XM_006228372.2 | XP_006228434.2 | 6, 7 | 469 | |||
XM_008758885.1 | XP_008757107.1 | 6, 7 | 535 | |||
GenBank mRNA | L25863.1 | - | 6, 7 | 420 |