Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Ctrl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_054009.1 | NP_446461.1 | 1 | 28 | ||
GenBank mRNA | AB020757.1 | - | 1 | 28 | ||
FQ234836.1 | - | 1 | 40 |